Assay Details
| Gene Symbol: IMMT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002757538.3 | XP_002757584.1 | 14 - 15 | 2358 | 114 | |
| XM_002757539.4 | XP_002757585.1 | 14 - 15 | 2395 | 114 | ||
| XM_008980384.2 | XP_008978632.1 | 14 - 15 | 2392 | 114 | ||
| XM_008980385.2 | XP_008978633.1 | 14 - 15 | 2361 | 114 | ||
| XM_008980386.2 | XP_008978634.1 | 14 - 15 | 2358 | 114 | ||
| XM_008980387.2 | XP_008978635.1 | 14 - 15 | 2356 | 114 | ||
| XM_008980388.2 | XP_008978636.1 | 13 - 14 | 2298 | 114 | ||
| XM_008980389.2 | XP_008978637.1 | 13 - 14 | 2264 | 114 | ||
| XM_008980390.2 | XP_008978638.1 | 13 - 14 | 2261 | 114 | ||
| XM_017964188.1 | XP_017819677.1 | 13 - 14 | 2384 | 114 | ||
More Information
Set Membership: |
|

IMMT
Genomic Map