Assay Details
| Gene Symbol: PTGES3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752617.4 | XP_002752663.2 | 5 - 6 | 787 | 99 | |
| XM_009004032.2 | XP_009002280.1 | 5 - 6 | 1094 | 99 | ||
| XM_009004033.2 | XP_009002281.1 | 5 - 6 | 1094 | 99 | ||
| XM_009004034.2 | XP_009002282.1 | 5 - 6 | 794 | 99 | ||
More Information
Set Membership: |
|

PTGES3
Genomic Map