Assay Details
| Gene Symbol: LOC100398454 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008987364.2 | XP_008985612.1 | 1 | 222 | 129 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100398454| Gene Symbol: LOC100398454 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008987364.2 | XP_008985612.1 | 1 | 222 | 129 | |
Set Membership: |
|