Assay Details
| Gene Symbol: SLC25A17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008979476.2 | XP_008977724.2 | 7 - 8 | 844 | 129 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SLC25A17| Gene Symbol: SLC25A17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008979476.2 | XP_008977724.2 | 7 - 8 | 844 | 129 | |
Set Membership: |
|