Assay Details
| Gene Symbol: TPM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008999047.2 | XP_008997295.1 | 8 - 9 | 1000 | 107 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TPM2| Gene Symbol: TPM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008999047.2 | XP_008997295.1 | 8 - 9 | 1000 | 107 | |
Set Membership: |
|