Assay Details
| Gene Symbol: LOC100387330 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008984425.2 | XP_008982673.1 | 2 - 3 | 261 | 114 | |
| XM_017966292.1 | XP_017821781.1 | 2 - 3 | 431 | 114 | ||
More Information
Set Membership: |
|

LOC100387330
Genomic Map