Assay Details
| Gene Symbol: ZMPSTE24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002750653.4 | XP_002750699.1 | 10 | 2827 | 72 | |
| XM_009000968.2 | XP_008999216.1 | 9 | 2683 | 72 | ||
More Information
Set Membership: |
|

ZMPSTE24
Genomic Map