Assay Details
| Gene Symbol: LOC100402247 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002764176.4 | XP_002764222.3 | 8 | 1216 | 149 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100402247| Gene Symbol: LOC100402247 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002764176.4 | XP_002764222.3 | 8 | 1216 | 149 | |
Set Membership: |
|