Assay Details
| Gene Symbol: RNF7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002759536.3 | XP_002759582.1 | 3 | 924 | 69 | |
| XM_008983734.2 | XP_008981982.1 | 3 | 920 | 69 | ||
More Information
Set Membership: |
|

RNF7
Genomic Map