Assay Details
| Gene Symbol: HSP90AA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002754299.4 | XP_002754345.2 | 4 | 822 | 115 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HSP90AA1| Gene Symbol: HSP90AA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002754299.4 | XP_002754345.2 | 4 | 822 | 115 | |
Set Membership: |
|