Assay Details
| Gene Symbol: SNRPG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002757650.4 | XP_002757696.1 | 4 | 522 | 108 | |
| XM_008980543.2 | XP_008978791.1 | 5 | 584 | 108 | ||
| XM_017964249.1 | XP_017819738.1 | 5 | 431 | 108 | ||
| XM_017964250.1 | XP_017819739.1 | 3 | 505 | 108 | ||
More Information
Set Membership: |
|

SNRPG
Genomic Map