Assay Details
| Gene Symbol: WDR6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002758328.3 | XP_002758374.3 | 6 | 4310 | 62 | |
| XM_017964895.1 | XP_017820384.1 | 6 | 3941 | 62 | ||
More Information
Set Membership: |
|

WDR6
Genomic Map