Assay Details
| Gene Symbol: SMARCE1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002748566.4 | XP_002748612.1 | 11 | 2222 | 119 | |
| XM_002748567.4 | XP_002748613.1 | 10 | 2117 | 119 | ||
| XM_008997534.2 | XP_008995782.1 | 9 | 2073 | 119 | ||
More Information
Set Membership: |
|

SMARCE1
Genomic Map