Assay Details
| Gene Symbol: B3GNT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002757714.3 | XP_002757760.1 | 2 | 1120 | 105 | |
| XM_017964285.1 | XP_017819774.1 | 3 | 1184 | 105 | ||
More Information
Set Membership: |
|

B3GNT2
Genomic Map