Assay Details
| Gene Symbol: PIP4K2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017976160.1 | XP_017831649.1 | 10 | 1512 | 70 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PIP4K2C| Gene Symbol: PIP4K2C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017976160.1 | XP_017831649.1 | 10 | 1512 | 70 | |
Set Membership: |
|