Assay Details
| Gene Symbol: LRIT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756166.3 | XP_002756212.1 | 4 | 2925 | 80 | |
| XM_002756167.3 | XP_002756213.1 | 3 | 2958 | 80 | ||
More Information
Set Membership: |
|

LRIT2
Genomic Map