Assay Details
| Gene Symbol: SKP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002744585.4 | XP_002744631.2 | 6 | 919 | 82 | |
| XM_008991210.2 | XP_008989458.1 | 7 | 946 | 82 | ||
More Information
Set Membership: |
|

SKP1
Genomic Map