Assay Details
Gene Symbol: NR2C2AP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002761926.4 | XP_002761972.1 | 5 | 1118 | 78 | |
XM_008987549.2 | XP_008985797.1 | 5 | 1125 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
