Assay Details
| Gene Symbol: NAP1L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752778.4 | XP_002752824.1 | 16 | 2815 | 154 | |
| XM_009004254.2 | XP_009002502.2 | 15 | 2413 | 154 | ||
| XM_009004255.2 | XP_009002503.2 | 15 | 2385 | 154 | ||
| XM_009004256.2 | XP_009002504.1 | 17 | 2564 | 154 | ||
| XM_009004258.2 | XP_009002506.1 | 14 | 2365 | 154 | ||
| XM_017976221.1 | XP_017831710.1 | 15 | 2486 | 154 | ||
| XM_017976222.1 | XP_017831711.1 | 15 | 2357 | 154 | ||
More Information
Set Membership: |
|

NAP1L1
Genomic Map