Assay Details
| Gene Symbol: Ccl5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172927.1 | NP_001166398.1 | 2 - 3 | 237 | 84 | |
| GenBank mRNA | AB002662.1 | - | 2 - 3 | 237 | 84 | |
| U77037.1 | - | 2 - 3 | 195 | 84 | ||
More Information
Set Membership: |
|

Ccl5
Genomic Map