Assay Details
| Gene Symbol: Ptgr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172980.1 | NP_001166451.1 | 5 - 6 | 537 | 67 | |
| GenBank mRNA | AB021219.1 | - | 5 - 6 | 537 | 67 | |
More Information
Set Membership: |
|

Ptgr1
Genomic Map