Assay Details
| Gene Symbol: Kcnj2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172975.1 | NP_001166446.1 | 2 | 639 | 89 | |
| XM_013142291.2 | XP_012997745.1 | 2 | 740 | 89 | ||
| GenBank mRNA | AF021142.1 | - | 1 | 355 | 89 | |
| AF183914.1 | - | 2 | 639 | 89 | ||
More Information
Set Membership: |
|

Kcnj2
Genomic Map