Assay Details
| Gene Symbol: Opn1mw | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172989.1 | NP_001166460.1 | 3 - 4 | 588 | 57 | |
| GenBank mRNA | AF132042.1 | - | 3 - 4 | 588 | 57 | |
More Information
Set Membership: |
|

Opn1mw
Genomic Map