Assay Details
| Gene Symbol: Hrh3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172737.1 | NP_001166208.1 | 1 - 2 | 420 | 95 | |
| GenBank mRNA | AF267537.1 | - | 1 - 2 | 420 | 95 | |
| AF267538.1 | - | 420 | 95 | |||
More Information
Set Membership: |
|

Hrh3
Genomic Map