Assay Details
| Gene Symbol: Ramp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173010.1 | NP_001166481.1 | 2 - 3 | 124 | 59 | |
| XM_013149406.1 | XP_013004860.1 | 2 - 3 | 689 | 59 | ||
| GenBank mRNA | AF484220.1 | - | 2 - 3 | 124 | 59 | |
More Information
Set Membership: |
|

Ramp2
Genomic Map