Assay Details
| Gene Symbol: Actb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172909.1 | NP_001166380.1 | 1 - 2 | 129 | 91 | |
| XM_005003653.3 | XP_005003710.1 | 2 - 3 | 231 | 91 | ||
| GenBank mRNA | AF508792.1 | - | 1 - 2 | 129 | 91 | |
More Information
Set Membership: |
|

Actb
Genomic Map