Assay Details
| Gene Symbol: Ppara | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173004.1 | NP_001166475.1 | 5 - 6 | 952 | 65 | |
| GenBank mRNA | AJ000222.1 | - | 5 - 6 | 952 | 65 | |
| AJ006218.1 | - | 5 - 6 | 918 | 65 | ||
More Information
Set Membership: |
|

Ppara
Genomic Map