Assay Details
| Gene Symbol: Adora2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172733.1 | NP_001166204.1 | 1 - 2 | 339 | 103 | |
| XM_005007205.3 | XP_005007262.1 | 2 - 3 | 768 | 103 | ||
| XM_013141514.2 | XP_012996968.1 | 2 - 3 | 661 | 103 | ||
| XM_013141515.1 | XP_012996969.1 | 2 - 3 | 647 | 103 | ||
| GenBank mRNA | D63674.1 | - | 1 - 2 | 356 | 103 | |
| U04201.1 | - | 1 - 2 | 339 | 103 | ||
More Information
Set Membership: |
|

Adora2a
Genomic Map