Assay Details
| Gene Symbol: Camp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173059.2 | NP_001166530.1 | 3 - 4 | 391 | 74 | |
| GenBank mRNA | D87405.1 | - | 3 - 4 | 392 | 74 | |
More Information
Set Membership: |
|

Camp
Genomic Map