Assay Details
| Gene Symbol: Ppy | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173005.1 | NP_001166476.1 | 2 - 3 | 197 | 57 | |
| GenBank mRNA | M29543.1 | - | 2 - 3 | 197 | 57 | |
More Information
Set Membership: |
|

Ppy
Genomic Map