Assay Details
| Gene Symbol: Serpina3k | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173013.1 | NP_001166484.1 | 1 - 2 | 625 | 71 | |
| XM_013144836.2 | XP_013000290.1 | 2 - 3 | 708 | 71 | ||
| GenBank mRNA | M57269.1 | - | 1 - 2 | 625 | 71 | |
| Gene Symbol: Apf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005000487.3 | XP_005000544.1 | 2 - 3 | 683 | 71 | |
| GenBank mRNA | M57271.1 | - | 1 - 2 | 605 | 71 | |
| Gene Symbol: Sh2b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172734.1 | NP_001166205.1 | 2 - 3 | 640 | 71 | |
| GenBank mRNA | M57270.1 | - | 2 - 3 | 640 | 71 | |
More Information
Set Membership: |
|

Serpina3k
Genomic Map