Assay Details
| Gene Symbol: Apoc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172914.2 | NP_001166385.1 | 1 - 2 | 80 | 96 | |
| XM_013146781.2 | XP_013002235.1 | 2 - 3 | 160 | 96 | ||
| GenBank mRNA | M59913.1 | - | 1 - 2 | 84 | 96 | |
More Information
Set Membership: |
|

Apoc2
Genomic Map