Assay Details
| Gene Symbol: Adora1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172911.1 | NP_001166382.1 | 1 - 2 | 848 | 84 | |
| XM_013157793.2 | XP_013013247.1 | 3 - 4 | 823 | 84 | ||
| XM_013157794.2 | XP_013013248.1 | 3 - 4 | 742 | 84 | ||
| XM_013157795.1 | XP_013013249.1 | 2 - 3 | 487 | 84 | ||
| XM_013157796.1 | XP_013013250.1 | 2 - 3 | 509 | 84 | ||
| XM_013157797.2 | XP_013013251.1 | 2 - 3 | 695 | 84 | ||
| XM_013157798.2 | XP_013013252.1 | 2 - 3 | 661 | 84 | ||
| GenBank mRNA | U04279.1 | - | 1 - 2 | 848 | 84 | |
More Information
Set Membership: |
|

Adora1
Genomic Map