Assay Details
| Gene Symbol: Kcnh2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172973.1 | NP_001166444.1 | 5 - 6 | 1577 | 75 | |
| GenBank mRNA | EF204534.1 | - | 5 - 6 | 1577 | 75 | |
| U75211.1 | - | 1 - 2 | 338 | 75 | ||
More Information
Set Membership: |
|

Kcnh2
Genomic Map