Assay Details
| Gene Symbol: Ocm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172988.2 | NP_001166459.1 | 4 - 5 | 376 | 95 | |
| GenBank mRNA | U77842.1 | - | 4 - 5 | 376 | 95 | |
More Information
Set Membership: |
|

Ocm2
Genomic Map