Assay Details
| Gene Symbol: Spam1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173021.2 | NP_001166492.1 | 2 - 3 | 1305 | 92 | |
| GenBank mRNA | X56332.1 | - | 2 - 3 | 1306 | 92 | |
More Information
Set Membership: |
|

Spam1
Genomic Map