Assay Details
| Gene Symbol: Pnlip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173001.1 | NP_001166472.1 | 5 - 6 | 651 | 65 | |
| GenBank mRNA | X77403.1 | - | 5 - 6 | 651 | 65 | |
More Information
Set Membership: |
|

Pnlip
Genomic Map