Assay Details
| Gene Symbol: Pla2g2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173000.1 | NP_001166471.1 | 2 - 3 | 89 | 92 | |
| GenBank mRNA | X82631.1 | - | 2 - 3 | 89 | 92 | |
More Information
Set Membership: |
|

Pla2g2a
Genomic Map