Assay Details
| Gene Symbol: LOC100135519 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172946.1 | NP_001166417.1 | 5 - 6 | 617 | 83 | |
| GenBank mRNA | X85112.1 | - | 5 - 6 | 617 | 83 | |
| X92742.1 | - | 5 - 6 | 617 | 83 | ||
More Information
Set Membership: |
|

LOC100135519
Genomic Map