Assay Details
| Gene Symbol: Ebp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172945.1 | NP_001166416.1 | 2 - 3 | 408 | 93 | |
| GenBank mRNA | Z37985.1 | - | 2 - 3 | 408 | 93 | |
More Information
Set Membership: |
|

Ebp
Genomic Map