Assay Details
| Gene Symbol: Cyp11b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172939.1 | NP_001166410.1 | 1203 | 70 | ||
| GenBank mRNA | Z69785.1 | - | 1203 | 70 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cyp11b1| Gene Symbol: Cyp11b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172939.1 | NP_001166410.1 | 1203 | 70 | ||
| GenBank mRNA | Z69785.1 | - | 1203 | 70 | ||
Set Membership: |
|