Assay Details
| Gene Symbol: Oprk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172990.1 | NP_001166461.1 | 3 | 1151 | 100 | |
| GenBank mRNA | U04092.1 | - | 3 | 1151 | 100 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Oprk1| Gene Symbol: Oprk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172990.1 | NP_001166461.1 | 3 | 1151 | 100 | |
| GenBank mRNA | U04092.1 | - | 3 | 1151 | 100 | |
Set Membership: |
|