Assay Details
| Gene Symbol: Calm3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173034.2 | NP_001166505.1 | 2 - 3 | 201 | 64 | |
| GenBank mRNA | FJ012165.1 | - | 2 - 3 | 66 | 64 | |
| HQ326646.1 | - | 2 - 3 | 201 | 64 | ||
More Information
Set Membership: |
|

Calm3
Genomic Map