Assay Details
| Gene Symbol: Fabp5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173092.1 | NP_001166563.1 | 1 - 2 | 134 | 81 | |
| GenBank mRNA | EB372154.1 | - | 1 - 2 | 134 | 81 | |
| EU264005.1 | - | 1 - 2 | 134 | 81 | ||
More Information
Set Membership: |
|

Fabp5
Genomic Map