Assay Details
| Gene Symbol: Lim2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173039.1 | NP_001166510.1 | 2 - 3 | 204 | 58 | |
| XM_013147647.1 | XP_013003101.1 | 1 - 2 | 478 | 58 | ||
| GenBank mRNA | EB370137.1 | - | 2 - 3 | 204 | 58 | |
| EU827600.1 | - | 2 - 3 | 204 | 58 | ||
More Information
Set Membership: |
|

Lim2
Genomic Map