Assay Details
| Gene Symbol: Lim2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173039.1 | NP_001166510.1 | 4 - 5 | 487 | 52 | |
| XM_013147647.1 | XP_013003101.1 | 3 - 4 | 761 | 52 | ||
| GenBank mRNA | EB370137.1 | - | 4 - 5 | 487 | 52 | |
| EU827600.1 | - | 4 - 5 | 487 | 52 | ||
More Information
Set Membership: |
|

Lim2
Genomic Map