Assay Details
| Gene Symbol: Coch | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173043.1 | NP_001166514.1 | 8 - 9 | 746 | 75 | |
| XM_005001381.3 | XP_005001438.2 | 9 - 10 | 846 | 75 | ||
| GenBank mRNA | EU881087.1 | - | 8 - 9 | 746 | 75 | |
More Information
Set Membership: |
|

Coch
Genomic Map