Assay Details
| Gene Symbol: Sybu | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001173111.1 | NP_001166582.1 | 2 - 3 | 379 | 78 | |
| XM_005008208.3 | XP_005008265.1 | 5 - 6 | 712 | 78 | ||
| XM_013143704.2 | XP_012999158.1 | 5 - 6 | 768 | 78 | ||
| XM_013143705.2 | XP_012999159.1 | 4 - 5 | 652 | 78 | ||
| XM_013143706.2 | XP_012999160.1 | 2 - 3 | 2488 | 78 | ||
| XM_013143707.1 | XP_012999161.1 | 3 - 4 | 259 | 78 | ||
| XM_013143708.2 | XP_012999162.1 | 5 - 6 | 712 | 78 | ||
| GenBank mRNA | AF494296.1 | - | 2 - 3 | 379 | 78 | |
More Information
Set Membership: |
|

Sybu
Genomic Map