Assay Details
| Gene Symbol: Cd1b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172850.1 | NP_001166321.1 | 1 - 2 | 65 | 66 | |
| GenBank mRNA | AF145484.1 | - | 1 - 2 | 65 | 66 | |
More Information
Set Membership: |
|

Cd1b2
Genomic Map