Assay Details
| Gene Symbol: Cd8b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001172877.1 | NP_001166348.1 | 4 - 5 | 639 | 100 | |
| GenBank mRNA | AY303774.1 | - | 4 - 5 | 639 | 100 | |
More Information
Set Membership: |
|

Cd8b
Genomic Map